MilliporeSigma
  • Utilisation of phytate phosphorus by rumen bacteria in a semi-continuous culture system (Rusitec) in lactating goats fed on different forage to concentrate ratios.
Utilisation of phytate phosphorus by rumen bacteria in a semi-continuous culture system (Rusitec) in lactating goats fed on different forage to concentrate ratios.
Reproduction, nutrition, development (2001-10-11)
S Godoy, F Meschy
ABSTRACT
Experimental data on phytate phosphorus utilisation by ruminants are scarce. The aim of this study was to estimate the phytase activity of rumen micro-organisms when phytate phosphorus supply is high. A semi-continuous culture system fermentor (RUSITEC) was used. The inoculum was obtained from eight goats fed on either high or low forage level diets. Experimental buffers only differed by the nature of phosphorus monosodium phosphate vs. corn sodium phytate. The nylon bags containing 15 g DM of substrate were removed after a 48-hour incubation period. The system was maintained for 15 days: 5 days for adaptation, in order to obtain a steady state, and 10 days for sampling and recording. No significant differences were observed for DM digestibility, gas production, pH, N-NH3, and SCFA for the different treatments. Bacterial efficiency of phytate phosphorus utilisation was significantly higher (p < 0.001) with organic P, but remained lower than the data usually reported in the literature. These results may be explained by the relative saturation of bacterial phytase activity when the buffer contains a high level of phytate phosphorus.
Sigma-Aldrich
Phytic acid sodium salt hydrate, from rice