  • Search Results
  • Distinct iNKT Cell Populations Use IFNγ or ER Stress-Induced IL-10 to Control Adipose Tissue Homeostasis.
Distinct iNKT Cell Populations Use IFNγ or ER Stress-Induced IL-10 to Control Adipose Tissue Homeostasis.
Cell metabolism (2020-06-10)
Nelson M LaMarche, Harry Kane, Ayano C Kohlgruber, Han Dong, Lydia Lynch, Michael B Brenner
ABSTRACT
Adipose tissue invariant natural killer T (iNKT) cells are phenotypically different from other iNKT cells because they produce IL-10 and control metabolic homeostasis. Why that is the case is unclear. Here, using single-cell RNA sequencing, we found several adipose iNKT clusters, which we grouped into two functional populations based on NK1.1 expression. NK1.1NEG cells almost exclusively produced IL-10 and other regulatory cytokines, while NK1.1POS iNKT cells predominantly produced IFNγ. Mechanistically, biochemical fractionation revealed that free fatty acids drive IL-10 production primarily in NK1.1NEG iNKT cells via the IRE1α-XBP1s arm of the unfolded protein response. Correspondingly, adoptive transfer of adipose tissue NK1.1NEG iNKT cells selectively restored metabolic function in obese mice. Further, we found an unexpected role for NK1.1POS iNKT cells in lean adipose tissue, as IFNγ licenses natural killer cell-mediated macrophage killing to limit pathological macrophage expansion. Together, these two iNKT cell populations utilize non-redundant pathways to preserve metabolic integrity.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
4μ8C, ≥98% (HPLC)
Sigma-Aldrich
Sodium phenylbutyrate, ≥98% (HPLC)
Sigma-Aldrich
Tunicamycin from Streptomyces sp.