  • Search Results
  • Synthesis and evaluation of cytotoxic effects of novel α-methylenelactone tetracyclic diterpenoids.
Synthesis and evaluation of cytotoxic effects of novel α-methylenelactone tetracyclic diterpenoids.
Bioorganic & medicinal chemistry letters (2012-02-10)
Yao-Fu Zeng, Jia-Qiang Wu, Lian-Yong Shi, Ke Wang, Bin Zhou, Yong Tang, Da-Yong Zhang, Yang-Chang Wu, Wei-Yi Hua, Xiao-Ming Wu
ABSTRACT
A series of tetracyclic diterpenoids bearing the α-methylenelactone group have been synthesized and screened for their in vitro anti-tumor activities against six human cancer cell lines. The results showed that compounds 1c, 2a and 2b exhibited significant cytotoxicity superior to the positive control doxorubicin hydrochloride against MDA-MB-231, K562 and HepG2 cell lines. In particular, compound 2b was identified as the most promising anticancer agent against HepG2 cells with IC(50) value of 0.09μM.
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)