  • Search Results
  • Solvent free synthesis, anti-inflammatory and anticancer activity evaluation of tricyclic and tetracyclic benzimidazole derivatives.
Solvent free synthesis, anti-inflammatory and anticancer activity evaluation of tricyclic and tetracyclic benzimidazole derivatives.
Bioorganic & medicinal chemistry letters (2010-03-02)
Sham M Sondhi, Reshma Rani, Jaiveer Singh, Partha Roy, S K Agrawal, A K Saxena
ABSTRACT
Heterocyclic benzimidazole derivatives 3a-h, 5a-c and 7a-d have been synthesized by condensation of succinic acid (1) homophthalic acid (4) and 2,3-pyrazinedicarboxlic acid (6) with various substituted diamines under microwave irradiation in good yields. Structures assigned to 3a-h, 5a-c and 7a-d are fully supported by spectral data. All these compounds were screened for anti-inflammatory and anticancer activities. At a dose of 50 mg/kg p.o. compounds 3b (39.4%) and 3c (39.2%) exhibited anti-inflammatory activity, comparable to standard ibuprofen which showed 39% activity at 50 mg/kg p.o. and compound 7c exhibit good anticancer activity against ovary (IGR-OV-1), breast (MCF-7) and CNS(SF-295) human cancer cell lines.
Product Description
Supelco
Ibuprofen
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥970 μg/mg (USP XXIV)