  • Search Results
  • Anti-inflammatory flavonoids from the rhizomes of Helminthostachys zeylanica.
Anti-inflammatory flavonoids from the rhizomes of Helminthostachys zeylanica.
Journal of natural products (2009-07-09)
Yaun-Chao Huang, Tsong-Long Hwang, Chih-Shiang Chang, Yu-Liang Yang, Chia-Ning Shen, Wen-Ying Liao, Sheng-Chih Chen, Chih-Chuang Liaw
ABSTRACT
Eight new prenylated flavonoids, ugonins M-T (1-8), together with five known compounds, ugonins J-L (9-11), 5,4'-dihydroxy-4'',4''-dimethyl-5''-methyl-5''H-dihydrofurano[2'',3'':6,7]flavanone, and quercetin, were isolated and purified from the rhizomes of Helminthostachys zeylanica. The structures of the new isolates were elucidated by spectroscopic and chemical methods. Compounds 1, 3, 5, 7, 8, and 11 showed inhibition of superoxide anion generation and elastase release by human neutrophils in response to formyl-l-methionyl-l-leucyl-l-phenylalanine/cytochalasin B (FMLP/CB).
Sigma-Aldrich
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)