  • Search Results
  • An artificial habitat increases the reproductive fitness of a range-shifting species within a newly colonized ecosystem.
An artificial habitat increases the reproductive fitness of a range-shifting species within a newly colonized ecosystem.
Scientific reports (2020-01-19)
Zachary J Cannizzo, Susan Q Lang, Bryan Benitez-Nelson, Blaine D Griffen
ABSTRACT
When a range-shifting species colonizes an ecosystem it has not previously inhabited, it may experience suboptimal conditions that challenge its continued persistence and expansion. Some impacts may be partially mitigated by artificial habitat analogues: artificial habitats that more closely resemble a species' historic ecosystem than the surrounding habitat. If conditions provided by such habitats increase reproductive success, they could be vital to the expansion and persistence of range-shifting species. We investigated the reproduction of the mangrove tree crab Aratus pisonii in its historic mangrove habitat, the suboptimal colonized salt marsh ecosystem, and on docks within the marsh, an artificial mangrove analogue. Crabs were assessed for offspring production and quality, as well as measures of maternal investment and egg quality. Aratus pisonii found on docks produced more eggs, more eggs per unit energy investment, and higher quality larvae than conspecifics in the surrounding salt marsh. Yet, crabs in the mangrove produced the highest quality larvae. Egg lipids suggest these different reproductive outcomes result from disparities in the quality of diet-driven maternal investments, particularly key fatty acids. This study suggests habitat analogues may increase the reproductive fitness of range-shifting species allowing more rapid expansion into, and better persistence in, colonized ecosystems.
Product Description
Supelco
Supelco 37 Component FAME Mix, certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL
Sigma-Aldrich
Glycogen Assay Kit, sufficient for 100 colorimetric or fluorometric tests