MilliporeSigma
Anti-tumor 12-deoxyphorbol esters from Euphorbia cornigera.
European journal of medicinal chemistry (2007-05-22)
Imam Bakhsh Baloch, Musa Kaleem Baloch, Qazi Najam us Saqib
ABSTRACT
Nine (1-8 and 10) new and two (9 and 11) known compounds have been isolated from roots of Euphorbia cornigera Boiss. Their structure and relative stereochemistry were acquired through NMR ((1)H, (13)C, COSY-45, HOHAHA, HMQC, HMBC, NOE and HMBC) spectroscopic measurements. Compounds 1-10 were identified as diesters of 13,20-O-diacyl and 11 as 13-O-acetyl of 12-deoxyphorbol. Cytotoxic activity of the compounds was investigated on human KB cells by reduction of MTT. Compounds 8-10 displayed IC(50) of 0.8, 0.5, and 1.0 microg mL(-1), respectively, whereas the activity of rest of the compounds (1-7) was either very low or (11) zero even up to 1000 microg mL(-1). The inhibition of DNA synthesis through Trypan blue exclusion and Brd-U assay was investigation to figure out the role of compounds 8-10 and concluded that these were responsible for the death of KB cells. Significant correlation has been found between the cytotoxicity and DNA cross-link and DNA strand-break formation.
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)