  • Search Results
  • Quercinol, an anti-inflammatory chromene from the wood-rotting fungus Daedalea quercina (Oak Mazegill).
Quercinol, an anti-inflammatory chromene from the wood-rotting fungus Daedalea quercina (Oak Mazegill).
Bioorganic & medicinal chemistry letters (2007-03-10)
P Gebhardt, K Dornberger, F A Gollmick, U Gräfe, A Härtl, H Görls, B Schlegel, C Hertweck
ABSTRACT
The fungus Daedalea quercina (oak mazegill) was examined for its capability of producing antioxidative and anti-inflammatory compounds. Bioactivity guided fractionation of the extract from a mycelial culture led to the isolation of quercinol, which was identified as (-)-(2S)-2-hydroxymethyl-2-methyl-6-hydroxychromene 1 by NMR and X-ray analyses. The cryptic hydroquinone 1 shows a broad anti-inflammatory activity against cyclooxygenase 2 (COX-2), xanthine oxidase (XO), and horseradish peroxidase (HRP) at micromolar concentrations.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)