  • Search Results
  • Formaldehyde-based whole-mount in situ hybridization method for planarians.
Formaldehyde-based whole-mount in situ hybridization method for planarians.
Developmental dynamics : an official publication of the American Association of Anatomists (2009-01-24)
Bret J Pearson, George T Eisenhoffer, Kyle A Gurley, Jochen C Rink, Diane E Miller, Alejandro Sánchez Alvarado
ABSTRACT
Whole-mount in situ hybridization (WISH) is a powerful tool for visualizing gene expression patterns in specific cell and tissue types. Each model organism presents its own unique set of challenges for achieving robust and reproducible staining with cellular resolution. Here, we describe a formaldehyde-based WISH method for the freshwater planarian Schmidtea mediterranea developed by systematically comparing and optimizing techniques for fixation, permeabilization, hybridization, and postprocessing. The new method gives robust, high-resolution labeling in fine anatomical detail, allows co-labeling with fluorescent probes, and is sufficiently sensitive to resolve the expression pattern of a microRNA in planarians. Our WISH methodology not only provides significant advancements over current protocols that make it a valuable asset for the planarian community, but should also find wide applicability in WISH methods used in other systems.
Product Description
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
TWEEN® 20, viscosity 250-450 mPa.s (25 °C)
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material