MilliporeSigma
CXCL10 blockade protects mice from cyclophosphamide-induced cystitis.
Journal of immune based therapies and vaccines (2008-10-30)
Senthilkumar K Sakthivel, Udai P Singh, Shailesh Singh, Dennis D Taub, Kristian R Novakovic, James W Lillard
ABSTRACT
Alterations in serum CXCR3 ligand levels were examined in interstitial cystitis (IC) patients; similar expression patterns in serum as well as CXCR3, CXCR3 ligands, and cytokines expressed by peripheral and local leukocyte subpopulations were characterized during cyclophosphamide (CYP)-induced acute cystitis in mice. Serum levels of monokine-induced by interferon-gamma (IFN-gamma) (MIG/CXCL9), IFN-gamma-inducible protein-10 (IP-10/CXCL10), and IFN-gamma-inducible T cell alpha chemoattractant (I-TAC/CXCL11) were elevated in patients with IC. These clinical features closely correlated with CYP-induced cystitis in mice. Serum levels of these CXCR3 ligands and local T helper type 1 (Th1) cytokines were also increased. We demonstrate that CXCR3 as well as CXCL9, CXCL10 and CXCL11 mRNA were significantly expressed by urinary bladder lymphocytes, while CXCR3 and CXCL9 transcripts were significantly expressed by iliac lymph node leukocytes following CYP treatment. We also show that the number of CD4+ T cells, mast cells, natural killer (NK) cells, and NKT cells were increased at systemic (spleen) and mucosal (urinary bladder and iliac lymph nodes) sites, following CYP-induced cystitis in mice. Importantly, CXCL10 blockade attenuated these increases caused by CYP. Antibody (Ab)-mediated inhibition of the most abundant serum CXCR3 ligand, CXCL10, in mice decreased the local production of CXCR3 ligands as well as Th1 cytokines expressed by local leukocytes, and lowered corresponding serum levels to reduce the severity of CYP-induced cystitis. The present study is among the first to demonstrate some of the cellular and molecular mechanisms of chemokines in cystitis and may represent new drug target for this disease.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Freund′s Adjuvant, Incomplete, liquid
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%