  • Search Results
  • Calcium triggers the dissociation of myosin-Va from ribosomes in ribonucleoprotein complexes.
Calcium triggers the dissociation of myosin-Va from ribosomes in ribonucleoprotein complexes.
FEBS letters (2020-05-16)
Lucía Canclini, Karina Cal, Camila Bardier, Paul Ruiz, John A Mercer, Aldo Calliari
ABSTRACT
The sorting of RNAs to specific regions of the cell for local translation represents an important mechanism directing protein distribution and cell compartmentalization. While significant progress has been made in understanding the mechanisms underlying the transport and localization of mRNAs, the mechanisms governing ribosome mobilization are less well understood. Ribosomes present in the cytoplasm of multiple cell types can form ribonucleoprotein complexes that also contain myosin-Va (Myo5a), a processive, actin-dependent molecular motor. Here, we report that Myo5a can be disassociated from ribosomes when ribonucleoprotein complexes are exposed to calcium, both in vitro and in vivo. We suggest that Myo5a may act as a molecular switch able to anchor or release ribosomes from the actin cytoskeleton in response to intracellular signaling.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ionomycin calcium salt from Streptomyces conglobatus, powder, ≥98% (HPLC)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2μm filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%