  • Search Results
  • Possible roles of parathyroid hormone, 1.25(OH)2D3, and fibroblast growth factor 23 on genes controlling calcium metabolism across different tissues of the laying hen.
Possible roles of parathyroid hormone, 1.25(OH)2D3, and fibroblast growth factor 23 on genes controlling calcium metabolism across different tissues of the laying hen.
Domestic animal endocrinology (2020-02-02)
A Gloux, N Le Roy, J Ezagal, N Même, C Hennequet-Antier, M L Piketty, D Prié, G Benzoni, J Gautron, Y Nys, A Narcy, M J Duclos
ABSTRACT
This study provides an integrative description of candidate gene expression across tissues involved in calcium (Ca) metabolism during the egg laying cycle, using the well-defined model of Ca supply as fine or coarse particles of calcium carbonate (CaCO3). Plasma and tissue samples were collected from hens at the peak of laying at 0 to 1, 9 to 10, and 18 to 19 h postovulation (PO). After mRNA preparation from the parathyroid gland, medullary bone, liver, kidney, duodenum, and jejunum, gene expressions were quantified using RT-qPCR. The highest levels of parathyroid hormone (PTH) mRNA in the parathyroid gland (P < 0.05), and of the active form of vitamin D3 1.25(OH)2D3 in the plasma (P < 0.01) were observed at 18 to 19 h PO. During this active phase of eggshell formation, bone resorption was attested to high levels of plasma inorganic phosphorus (iP) and the receptor activation of nuclear factor-κB expression in the bone (P < 0.001 and P < 0.05, respectively). At this stage, 5 genes of the transcellular and the paracellular Ca absorption pathways in the intestine (P < 0.05) and the Ca channel transient receptor potential cation channel subfamily V member 5 (P < 0.05), involved in its reabsorption in the kidney, were overexpressed. At 0 to 1 h PO during the subsequent daylight period, 2 candidates of the transcellular and the paracellular Ca pathways (P < 0.05) remained at high levels in the intestine, while calbindin D 28K expression was the highest in the kidney (P < 0.05). As PTH mRNA and 1.25(OH)2D3 were low, bone accretion was likely active at this stage. The phosphaturic hormone fibroblast growth factor 23 (FGF23) was overexpressed at 18 to 19 h PO (P < 0.05) in the bone when plasma iP was high, which suggested a role in the subsequent reduction of P reabsorption in the kidney, as attested to the decreased expression of P cotransporters, leading to iP clearance from the plasma at 0 to 1 h PO (P < 0.05). The low levels of 1.25(OH)2D3 at this stage coincided with increased expression of the 24-hydroxylase gene in the kidney (P < 0.05). In hens fed fine particles of CaCO3, higher plasma levels of 1,25(OH)2D3 and higher expression of several genes involved in bone turnover reflected a stronger challenge to Ca homeostasis. Altogether, these data support the hypothesis that FGF23 could drive vitamin D metabolism in the laying hen, as previously documented in other species and explain the tight link between P and Ca metabolisms.
Sigma-Aldrich
TRI Reagent®, LS, For processing fluid samples such as cell suspensions, CSF, and amniotic fluid.
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets