  • Search Results
  • Restoration of HDAC2 and Nrf2 by andrographolide overcomes corticosteroid resistance in chronic obstructive pulmonary disease.
Restoration of HDAC2 and Nrf2 by andrographolide overcomes corticosteroid resistance in chronic obstructive pulmonary disease.
British journal of pharmacology (2020-04-27)
Wupeng Liao, Albert Y H Lim, W S Daniel Tan, John Abisheganaden, W S Fred Wong
ABSTRACT
Corticosteroid resistance poses a major barrier to an effective anti-inflammatory therapy for chronic obstructive pulmonary disease (COPD). The present study aimed to investigate potential corticosteroid re-sensitization actions of andrographolide, a bioactive molecule from the herb Andrographis paniculata, in COPD models, particularly in peripheral blood mononuclear cells (PBMCs) from COPD patients. Corticosteroid sensitivity in PBMCs collected from COPD patients, or in human monocytic U937 cells exposed to cigarette smoke extract (CSE), was determined by measuring LPS-induced IL-8 production, in the presence and absence of andrographolide. The mechanisms of corticosteroid re-sensitization action of andrographolide were evaluated in a mouse cigarette smoke (CS)-induced acute lung injury model. Impaired inhibition of IL-8 production by dexamethasone was detected in PBMCs from COPD patients and in CSE-exposed U937 cells, together with reduced levels of nuclear factor erythroid 2-related factor 2 (Nrf2) and histone deacetylase-2 (HDAC2). In both PBMCs and CSE-exposed U937 cells, andrographolide restored dexamethasone inhibition of IL-8 production, accompanied by the up-regulation of Nrf2 and HDAC2 levels. In the U937 cells, andrographolide was able to block CSE-induced Akt and reduce the level of c-Jun. Besides, andrographolide also augmented dexamethasone actions on lowering total and neutrophil counts, cytokine levels, and oxidative damage markers in bronchoalveolar lavage fluid from CS-exposed mice. We report here for the first time a novel corticosteroid re-sensitization property of andrographolide in human PBMCs and provide mechanistic evidence to support clinical evaluation of andrographolide in reversing steroid resistance in COPD.
Sigma-Aldrich
Dexamethasone 21-phosphate disodium salt, ≥98%