  • Search Results
  • Combining probiotic Phaeobacter inhibens DSM17395 and broad-host-range vibriophage KVP40 against fish pathogenic vibrios
Combining probiotic Phaeobacter inhibens DSM17395 and broad-host-range vibriophage KVP40 against fish pathogenic vibrios
Aquaculture (Amsterdam, Netherlands) (2019)
Rasmussen BB, et al