MilliporeSigma
Long-Term Fipronil Treatment Induces Hyperactivity in Female Mice.
International journal of environmental research and public health (2020-03-04)
Svenja Koslowski, Camille Latapy, Pierrïck Auvray, Marc Blondel, Laurent Meijer
ABSTRACT
Fipronil is an insecticide widely used for veterinary and agricultural purposes. While its insecticidal properties mostly rely on its high affinity antagonistic activity on insect γ aminobutyric acid (GABA) receptors, fipronil and its main metabolite fipronil sulfone nevertheless display non-negligible affinity for mammalian GABAA receptor. As several environmental toxicants have been shown to raise the risk of developing various neurodegenerative disorders, the aim of this study was to evaluate whether long-term low dose administration of fipronil could lead to cognitive deficiencies. Our results indicate that long-term fipronil treatment leads to behavioral perturbations in mice, indicating an accumulative effect of sustained exposure to low dose of fipronil. Although no memory impairment was observed during the course of our study, we noticed a significant hyperlocomotion behavior after 43 weeks of weekly fipronil administration, which is consistent with its direct effect on the GABAergic system.
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%