  • Search Results
  • Metal?Organic Semiconductor Nanostructures for Surface-Enhanced Raman Scattering
Metal?Organic Semiconductor Nanostructures for Surface-Enhanced Raman Scattering
Applied Spectroscopy (2011)
Alessio P, et al.