  • Search Results
  • Simultaneous preparation of SERS-active metal colloids and plates by laser ablation
Simultaneous preparation of SERS-active metal colloids and plates by laser ablation
Journal of Raman Spectroscopy (2001)
Lee I, et al.