  • Search Results
  • Immunoglobulin G from breast cancer patients in stage I stimulates muscarinic acetylcholine receptors in MCF7 cells and induces proliferation. Participation of nitric oxide synthase-derived nitric oxide.
Immunoglobulin G from breast cancer patients in stage I stimulates muscarinic acetylcholine receptors in MCF7 cells and induces proliferation. Participation of nitric oxide synthase-derived nitric oxide.
Journal of clinical immunology (2010-02-17)
María Pía Negroni, Gabriel L Fiszman, María E Azar, Carlos Cresta Morgado, Alejandro J Español, Laura T Pelegrina, Eulalia de la Torre, María Elena Sales
ABSTRACT
Muscarinic acetylcholine receptors (mAChR) belong to the G-protein-coupled receptor family and are extensively expressed in most cells in mammals. We had reported the expression of mAChR in murine and human breast tumors. The presence of antibodies in the sera of patients with different tumors directed against self-proteins has been recently described. In this work, we investigated the presence of autoantibodies against mAChR in the sera of breast cancer patients in stage I (T1N0Mx-IgG). IgG purification was performed by affinity chromatography in protein G-agarose. We also studied the ability of these antibodies to modulate the proliferation of MCF-7 breast tumor cells by the MTS colorimetric assay. The ability of T1N0Mx-IgG to stimulate muscarinic signaling pathway via nitric oxide synthase was tested by Griess reaction. We demonstrated M(3) and M(4) receptors expression in MCF-7 cells. T1N0Mx-IgG promotes cell proliferation, mimicking the action of the muscarinic agonist carbachol. This effect was preferentially due to M(3) receptor activation in tumor cells via phospholipase C-induced nitric oxide liberation by calcium-dependent nitric oxide synthases. IgG from control patients was unable to produce this effect. IgG from patients with breast cancer in early stages could be promoting tumor progression by muscarinic activation, and its presence could be determining the prognosis of this illness.
Product Description
Millipore
Protein G-Agarose, Fast Flow from Streptococcus sp., saline suspension
Millipore
Protein G–Agarose, lyophilized powder, Contains lactose stabilizers that must be removed prior to use.