MilliporeSigma
  • IL-34 causes inflammation and beta cell apoptosis and dysfunction in gestational diabetes mellitus.
IL-34 causes inflammation and beta cell apoptosis and dysfunction in gestational diabetes mellitus.
Endocrine connections (2019-10-28)
Chenghao Piao, Xiaojie Wang, Shiqiao Peng, Xinyu Guo, Hui Zhao, Li He, Yan Zeng, Fan Zhang, Kewen Zhu, Yiwei Wang
ABSTRACT
Gestational diabetes mellitus (GDM) is characterized by glucose intolerance during gestation. It is associated with a series of maternal and foetal complications. Interleukin (IL)-34 is a recently discovered pro-inflammatory cytokine that functions as a ligand for colony-stimulating factor-1 receptor (CSF-1R). The contribution of IL-34 in the development of multiple chronic inflammatory diseases and autoimmune diseases has been recently discovered. The aim of this study was to evaluate whether IL-34 participates in the pathogenesis of GDM. A total of 120 women were enrolled in this study, which included 60 GDM patients and age- and sex-matched healthy pregnant women. The expression of IL-34 in serum, cord blood and placental tissues was analysed by ELISA and Western blot assays. The association between IL-34 levels and clinical features was also studied. We additionally evaluated the effect of recombinant mouse IL-34 (rmIL-34) on apoptosis and pancreatic β cell function. We found that IL-34 expression is highly increased in serum, cord blood and placental tissues in patients with GDM. In addition, there was a positive association between serum IL-34 and insulin resistance and glucose concentrations. Our data also revealed that IL-34 contributes to the apoptosis of pancreatic β cells in GDM caused by CSF-1R. Furthermore, functional studies found that IL-34 inhibited pancreatic β cell function and cell viability, while CSF-1R inhibitor blocked this effect. IL-34 plays a crucial role in the development of GDM by targeting CSF-1R, insulin production and β cell function.
Sigma-Aldrich
Human IL34 / Interleukin-34 ELISA Kit