  • Search Results
  • An RNA-Binding Protein Secreted by a Bacterial Pathogen Modulates RIG-I Signaling.
An RNA-Binding Protein Secreted by a Bacterial Pathogen Modulates RIG-I Signaling.
Cell host & microbe (2019-11-26)
Alessandro Pagliuso, To Nam Tham, Eric Allemand, Stevens Robertin, Bruno Dupuy, Quentin Bertrand, Christophe Bécavin, Mikael Koutero, Valérie Najburg, Marie-Anne Nahori, Frédéric Tangy, Fabrizia Stavru, Sergey Bessonov, Andréa Dessen, Christian Muchardt, Alice Lebreton, Anastassia V Komarova, Pascale Cossart
ABSTRACT
RNA-binding proteins (RBPs) perform key cellular activities by controlling the function of bound RNAs. The widely held assumption that RBPs are strictly intracellular has been challenged by the discovery of secreted RBPs. However, extracellular RBPs have been described in eukaryotes, while secreted bacterial RBPs have not been reported. Here, we show that the bacterial pathogen Listeria monocytogenes secretes a small RBP that we named Zea. We show that Zea binds a subset of L. monocytogenes RNAs, causing their accumulation in the extracellular medium. Furthermore, during L. monocytogenes infection, Zea binds RIG-I, the non-self-RNA innate immunity sensor, potentiating interferon-β production. Mouse infection studies reveal that Zea affects L. monocytogenes virulence. Together, our results unveil that bacterial RNAs can be present extracellularly in association with RBPs, acting as "social RNAs" to trigger a host response during infection.
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
TRI Reagent®, LS, For processing fluid samples such as cell suspensions, CSF, and amniotic fluid.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
Spautin-1, ≥98% (HPLC)
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Anti-RLN3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution