  • Search Results
  • Antitumour effects of metformin and curcumin in human papillomavirus positive and negative head and neck cancer cells.
Antitumour effects of metformin and curcumin in human papillomavirus positive and negative head and neck cancer cells.
Molecular carcinogenesis (2019-07-25)
Cameron Lindsay, Morris Kostiuk, Dustin Conrad, Daniel A O'Connell, Jeffrey Harris, Hadi Seikaly, Vincent L Biron
ABSTRACT
The incidence of oropharyngeal squamous cell carcinoma (OPSCC) has significantly increased in recent decades due to human papillomavirus (HPV)-mediated oncogenesis. Unfortunately, a growing number of HPV-positive (+) OPSCC survivors are living with the irreversible side effects of treatment. The novel, well-tolerated chemotherapeutics with improved side effect profiles are, therefore, in high demand. Metformin is one such drug, widely used as a first-line oral agent in the treatment of type 2 diabetes mellitus. Curcumin is another well-tolerated agent quickly gaining attention for its medicinal properties. Both metformin and curcumin have been shown to display anticancer properties. This study aimed to determine the antitumor effects of these agents, individually and combined, in HPV+​​​​ ​​​and HPV-negative (-) head and neck squamous cell carcinoma (HNSCC) cell lines. This was achieved by assessing the efficacy of varying drug concentrations on the overall cell viability, proliferation, and expression of common HNSCC biomarkers. The results from protein and RNA expression data are highly variable, as expected, with multiple pathways being affected in cancer. 3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assays and immunofluorescence microscopy suggest that both agents are capable of slowing proliferation and inducing apoptosis. We conclude that curcumin and metformin display effective antitumor effects in both HPV+ and HPV- HNSCC cell lines. The curcumin effects appear more pronounced in the HPV- cell lines. Metformin appears to be more effective at reducing the overall cell numbers in HPV+ cell lines. Metformin and curcumin combined did not appear to have synergistic effects on the proliferation or apoptosis of the treated cell lines.
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture