  • Search Results
  • Analysis of fatty acids in mouse tissue via in situ transmethylation with biochar.
Analysis of fatty acids in mouse tissue via in situ transmethylation with biochar.
Environmental geochemistry and health (2017-04-30)
Jechan Lee, Yiu Fai Tsang, Jeong-Ik Oh, Seokmann Hong, Changsung Kim, Eilhann E Kwon
ABSTRACT
Lipid derivatization technology-mediated fatty acid profiling studies have been suggested to dissect the contents of lipids in white fat and brown fat tissue. The focus of this study is to profile fatty acid lipidomics in brown adipose tissue and white adipose tissue of mice by derivatizing their lipids into fatty acid methyl esters via in situ transmethylation using a rice husk-derived biochar as porous media. The in situ transmethylation using biochar is advantageous in biological analysis because there was no loss of samples inevitably occurring in the loss of lipid in solvent extraction and purification steps.
Product Description
Supelco
Supelco 37 Component FAME Mix, certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL