  • Search Results
  • Hsp90 Chaperones Bluetongue Virus Proteins and Prevents Proteasomal Degradation.
Hsp90 Chaperones Bluetongue Virus Proteins and Prevents Proteasomal Degradation.
Journal of virology (2019-08-04)
Bjorn-Patrick Mohl, Polly Roy
ABSTRACT
The molecular chaperone machinery is important for the maintenance of protein homeostasis within the cells. The principle activities of the chaperone machinery are to facilitate protein folding and organize conformationally dynamic client proteins. Prominent among the members of the chaperone family are heat shock protein 70 (Hsp70) and 90 (Hsp90). Like cellular proteins, viral proteins depend upon molecular chaperones to mediate their stabilization and folding. Bluetongue virus (BTV), which is a model system for the Reoviridae family, is a nonenveloped arbovirus that causes hemorrhagic disease in ruminants. This constitutes a significant burden upon animals of commercial significance, such as sheep and cattle. Here, for the first time, we examined the role of chaperone proteins in the viral lifecycle of BTV. Using a combination of molecular, biochemical, and microscopic techniques, we examined the function of Hsp90 and its relevance to BTV replication. We demonstrate that Hsp70, the chaperone that is commonly usurped by viral proteins, does not influence virus replication, while Hsp90 activity is important for virus replication by stabilizing BTV proteins and preventing their degradation via the ubiquitin-proteasome pathway. To our knowledge this is the first report showing the involvement of Hsp90 as a modulator of BTV infection.IMPORTANCE Protein chaperones are instrumental for maintaining protein homeostasis, enabling correct protein folding and organization; prominent members include heat shock proteins 70 and 90. Virus infections place a large burden on this homeostasis. Identifying and understanding the underlying mechanisms that facilitate Bluetongue virus replication and spread through the usurpation of host factors is of primary importance for the development of intervention strategies. Our data identify and show that heat shock protein 90, but not heat shock protein 70, stabilizes bluetongue virus proteins, safeguarding them from proteasomal degradation.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, IgG fraction of antiserum
Sigma-Aldrich
Anti-Guinea Pig IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Millipore
Protein A-Sepharose® from Staphylococcus aureus, lyophilized powder
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Green Fluorescent Protein (GFP) antibody, Mouse monoclonal, clone GSN149, purified from hybridoma cell culture