MilliporeSigma
  • Structural Basis of the Differential Binding of Engineered Knottins to Integrins αVβ3 and α5β1.
Structural Basis of the Differential Binding of Engineered Knottins to Integrins αVβ3 and α5β1.
Structure (London, England : 1993) (2019-07-30)
Johannes F Van Agthoven, Hengameh Shams, Frank V Cochran, José L Alonso, James R Kintzing, Kiavash Garakani, Brian D Adair, Jian-Ping Xiong, Mohammad R K Mofrad, Jennifer R Cochran, M Amin Arnaout
ABSTRACT
Targeting both integrins αVβ3 and α5β1 simultaneously appears to be more effective in cancer therapy than targeting each one alone. The structural requirements for bispecific binding of ligand to integrins have not been fully elucidated. RGD-containing knottin 2.5F binds selectively to αVβ3 and α5β1, whereas knottin 2.5D is αVβ3 specific. To elucidate the structural basis of this selectivity, we determined the structures of 2.5F and 2.5D as apo proteins and in complex with αVβ3, and compared their interactions with integrins using molecular dynamics simulations. These studies show that 2.5D engages αVβ3 by an induced fit, but conformational selection of a flexible RGD loop accounts for high-affinity selective binding of 2.5F to both integrins. The contrasting binding of the highly flexible low-affinity linear RGD peptides to multiple integrins suggests that a "Goldilocks zone" of conformational flexibility of the RGD loop in 2.5F underlies its selective binding promiscuity to integrins.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Millipore
Protein A-Sepharose® from Staphylococcus aureus, lyophilized powder
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse