  • Search Results
  • Quantitative simultaneous estimation of water-soluble vitamins, Riboflavin, Pyridoxine, Cyanocobalamin and Folic Acid in neutraceutical products by HPLC
Quantitative simultaneous estimation of water-soluble vitamins, Riboflavin, Pyridoxine, Cyanocobalamin and Folic Acid in neutraceutical products by HPLC
The Open Analytical Chemistry Journal (2009)
Perveen S, et al.