  • Search Results
  • Silver nanowires: Synthesis technologies, growth mechanism and multifunctional applications
Silver nanowires: Synthesis technologies, growth mechanism and multifunctional applications
Materials Science and Engineering, B (2017)
Zhang P, et al.