  • Search Results
  • Highly stretchable and sensitive strain sensor based on silver nanowire-elastomer nanocomposite
Highly stretchable and sensitive strain sensor based on silver nanowire-elastomer nanocomposite
ACS Nano (2014)
Amjadi M, et al.