  • Search Results
  • Selective synthesis of 3-hydroxy acids from Meldrum?s acids using SmI2-H2O.
Selective synthesis of 3-hydroxy acids from Meldrum?s acids using SmI2-H2O.
Nature Protocols (2012)
Szostak M, et al.