  • Search Results
  • Cardiac Risk of Noncardiac Surgery After Percutaneous Coronary Intervention With Second-Generation Drug-Eluting Stents.
Cardiac Risk of Noncardiac Surgery After Percutaneous Coronary Intervention With Second-Generation Drug-Eluting Stents.
Anesthesia and analgesia (2018-09-01)
Bradford B Smith, Matthew A Warner, Nafisseh S Warner, Andrew C Hanson, Mark M Smith, Charanjit S Rihal, Rajiv Gulati, Malcolm R Bell, Gregory A Nuttall
ABSTRACT
Noncardiac surgery (NCS) following percutaneous coronary intervention (PCI) with stenting is sometimes associated with major adverse cardiac events (MACEs). Second-generation drug-eluting stents (DES) were developed to decrease the incidence of MACE seen with bare metal and first-generation DES. The medical records of all adult patients who underwent second-generation DES placement between July 29, 2008 and July 28, 2011 followed by NCS between September 22, 2008 and July 1, 2013 were reviewed. All episodes of MACE following surgery were recorded. A total of 282 patients (74.8% male) were identified who underwent NCS after PCI with second-generation DES. MACE occurred in 15 patients (5.3%), including 11 deaths. The incidence of MACE changed significantly with time from PCI to NCS: 17.1%, 10.0%, 0.0%, and 3.1% for patients undergoing NCS at 0-90, 91-180, 181-365, and ≥366 days, respectively. Compared with those having NCS ≥366 days after PCI, the odds ratio for MACE (95% confidence interval) was 6.4 (1.9 to 21.3) at 0-90 days and 3.4 (0.8 to 15.3) at 91-180 days. Seven days prior to NCS, 146 (52%) patients were on dual antiplatelet therapy (DAPT), 106 (38%) were on aspirin, and 30 (11%) did not receive antiplatelet therapy. Excessive surgical bleeding occurred in 19 cases (6.7%). While observed bleeding rates were lowest in those not receiving antiplatelet therapy, there were no statistically significant differences based on the presence or absence of antiplatelet therapy (3% [1/30] for no antiplatelet therapy compared to 6% [6/106] for aspirin monotherapy and 8% [12/146] for DAPT; Fisher exact test: P = .655). The incidence of MACE in patients with second-generation DES undergoing NCS was 5.3% and was highest in the first 180 days following DES implantation. The rate of excessive surgical bleeding was 6.7% with the highest observed rate in those on DAPT. However, differences by the presence or absence of antiplatelet therapy were not significant, and future large observational studies will be necessary to further define bleeding risk with continued DAPT.
Sigma-Aldrich
N-Chlorosuccinimide, 98%