MilliporeSigma
Alkali Blues: Blue-Emissive Alkali Metal Pyrrolates.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-02-21)
Oliver Back, Christoph Förster, Thomas Basché, Katja Heinze
ABSTRACT
2-Iminopyrroles [HtBu L, 4-tert-butyl phenyl(pyrrol-2-ylmethylene)amine] are non-fluorescent π systems. However, they display blue fluorescence after deprotonation with alkali metal bases in the solid state and in solution at room temperature. In the solid state, the alkali metal 2-imino pyrrolates, M(tBu L), aggregate to dimers, [M(tBu L)(NCR)]2 (M=Li, R=CH3 , CH(CH3 )CNH2 ), or polymers, [M(tBu L)]n (M=Na, K). In solution (solv=CH3 CN, DMSO, THF, and toluene), solvated, uncharged monomeric species M(tBu L)(solv)m with N,N'-chelated alkali metal ions are present. Due to the electron-rich pyrrolate and the electron-poor arylimino moiety, the M(tBu L) chromophore possesses a low-energy intraligand charge-transfer (ILCT) excited state. The chelated alkali cations rigidify the chromophore, restricting intramolecular motions (RIM) by the chelation-enhanced fluorescence (CHEF) effect in solution and, consequently, switch-on a blue fluorescence emission.
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide, 95%