  • Search Results
  • Triacylglycerol Composition of Breast Milk during Different Lactation Stages.
Triacylglycerol Composition of Breast Milk during Different Lactation Stages.
Journal of agricultural and food chemistry (2019-02-02)
Tinglan Yuan, Ce Qi, Xinyue Dai, Yuan Xia, Cong Sun, Jin Sun, Renqiang Yu, Qin Zhou, Qingzhe Jin, Wei Wei, Xingguo Wang
ABSTRACT
Triacylglycerol (TAG) composition of breast milk plays an important role in improving digestion, absorption, and metabolism when consumed by infants. This study characterized the TAG profile of human colostrum, transitional, and mature milk samples from 103 women. Significant differences in the TAGs composition of breast milk fat from three lactation stages were observed. The TAGs with high molecular weight and unsaturated fatty acid (such as 1,3-olein-2-palmitin (OPO) and 1(3)-olein-2-palmitin-3(1)-linolein (OPL)) were enriched in colostrum, while the TAGs containing medium-chain fatty acids were more abundant in transitional and mature milk than that in colostrum. Of note, OPL was the most common TAG in breast milk of Chinese women while the most common TAG in breast milk of Western women was OPO. This data will promote the development of infant formulas in terms of the TAG composition more suitable for infants.
Product Description
Tripalmitin, NIST® SRM® 1595
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Triolein, European Pharmacopoeia (EP) Reference Standard