  • Search Results
  • Picropodophyllin inhibits type I endometrial cancer cell proliferation via disruption of the PI3K/Akt pathway.
Picropodophyllin inhibits type I endometrial cancer cell proliferation via disruption of the PI3K/Akt pathway.
Acta biochimica et biophysica Sinica (2019-06-07)
Lin Dong, Meirong Du, Qianzhou Lv
ABSTRACT
The type-I insulin-like growth factor receptor (IGF-IR) is overexpressed in endometrial cancer. High IGF-IR expression was considered as an important prognostic factor for tumor progression. The purpose of this study was to investigate the role and molecular mechanism of IGF-IR inhibitor picropodophyllin (PPP) in the growth and development of endometrial cancer. High expression of IGF-IR was observed in endometrial cancer tissues, as well as in ECC-1 and KLE cell lines. PPP suppressed the number of clones of ECC-1 and KLE cell lines; however, it had no significant effect on HEC-1-A cell line, which expressed lower IGF-IR than ECC-1 and KLE cell lines. Furthermore, PPP reduced cell proliferation capacity, inhibited the IGF-IR mRNA expression, and suppressed protein phosphorylation of IGF-IR and Akt in the three cell lines. In addition, PPP inhibited the protein expression of survivin in KLE cell line after 1 h of exposure, though this effect did not last for prolonged time. In conclusion, IGF-IR was mostly overexpressed in type I endometrial cancer. High IGF-IR expression was an important prognostic factor of tumor progression. PPP mediated the down-regulation of IGF-IR phosphorylation and inhibited cell proliferation via the PI3K/Akt signal pathway. PPP may have the potential to become a clinical treatment target in endometrial carcinoma.
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder