  • Search Results
  • Lung cancer deficient in the tumor suppressor GATA4 is sensitive to TGFBR1 inhibition.
Lung cancer deficient in the tumor suppressor GATA4 is sensitive to TGFBR1 inhibition.
Nature communications (2019-04-12)
Lei Gao, Yong Hu, Yahui Tian, Zhenzhen Fan, Kun Wang, Hongdan Li, Qian Zhou, Guandi Zeng, Xin Hu, Lei Yu, Shiyu Zhou, Xinyuan Tong, Hsinyi Huang, Haiquan Chen, Qingsong Liu, Wanting Liu, Gong Zhang, Musheng Zeng, Guangbiao Zhou, Qingyu He, Hongbin Ji, Liang Chen
ABSTRACT
Lung cancer is the leading cause of cancer-related deaths worldwide. Tumor suppressor genes remain to be systemically identified for lung cancer. Through the genome-wide screening of tumor-suppressive transcription factors, we demonstrate here that GATA4 functions as an essential tumor suppressor in lung cancer in vitro and in vivo. Ectopic GATA4 expression results in lung cancer cell senescence. Mechanistically, GATA4 upregulates multiple miRNAs targeting TGFB2 mRNA and causes ensuing WNT7B downregulation and eventually triggers cell senescence. Decreased GATA4 level in clinical specimens negatively correlates with WNT7B or TGF-β2 level and is significantly associated with poor prognosis. TGFBR1 inhibitors show synergy with existing therapeutics in treating GATA4-deficient lung cancers in genetically engineered mouse model as well as patient-derived xenograft (PDX) mouse models. Collectively, our work demonstrates that GATA4 functions as a tumor suppressor in lung cancer and targeting the TGF-β signaling provides a potential way for the treatment of GATA4-deficient lung cancer.
Sigma-Aldrich
Normal Mouse IgG, This Normal Mouse IgG is validated for use in ELISA, Flow Cytometry, Immunoblotting, Immunofluorescence, Immunohistochemistry, Immunoprecipitation for the detection of Mouse IgG, Non-immune.