  • Search Results
  • Therapeutic Monoclonal Antibodies for Ebola Virus Infection Derived from Vaccinated Humans.
Therapeutic Monoclonal Antibodies for Ebola Virus Infection Derived from Vaccinated Humans.
Cell reports (2019-04-04)
Pramila Rijal, Sean C Elias, Samara Rosendo Machado, Julie Xiao, Lisa Schimanski, Victoria O'Dowd, Terry Baker, Emily Barry, Simon C Mendelsohn, Catherine J Cherry, Jing Jin, Geneviève M Labbé, Francesca R Donnellan, Tommy Rampling, Stuart Dowall, Emma Rayner, Stephen Findlay-Wilson, Miles Carroll, Jia Guo, Xiao-Ning Xu, Kuan-Ying A Huang, Ayato Takada, Gillian Burgess, David McMillan, Andy Popplewell, Daniel J Lightwood, Simon J Draper, Alain R Townsend
ABSTRACT
We describe therapeutic monoclonal antibodies isolated from human volunteers vaccinated with recombinant adenovirus expressing Ebola virus glycoprotein (EBOV GP) and boosted with modified vaccinia virus Ankara. Among 82 antibodies isolated from peripheral blood B cells, almost half neutralized GP pseudotyped influenza virus. The antibody response was diverse in gene usage and epitope recognition. Although close to germline in sequence, neutralizing antibodies with binding affinities in the nano- to pico-molar range, similar to "affinity matured" antibodies from convalescent donors, were found. They recognized the mucin-like domain, glycan cap, receptor binding region, and the base of the glycoprotein. A cross-reactive cocktail of four antibodies, targeting the latter three non-overlapping epitopes, given on day 3 of EBOV infection, completely protected guinea pigs. This study highlights the value of experimental vaccine trials as a rich source of therapeutic human monoclonal antibodies.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
ExtrAvidin®−FITC, buffered aqueous solution
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Thermolysin from Geobacillus stearothermophilus, Type X, lyophilized powder, 30-350 units/mg protein (E1%/280)
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Millipore
Protein A-Sepharose® from Staphylococcus aureus, lyophilized powder
Sigma-Aldrich
SIGMAFAST OPD, tablet