  • Search Results
  • Inhibition of Eph/ephrin interaction with the small molecule UniPR500 improves glucose tolerance in healthy and insulin-resistant mice.
Inhibition of Eph/ephrin interaction with the small molecule UniPR500 improves glucose tolerance in healthy and insulin-resistant mice.
Pharmacological research (2019-01-10)
C Giorgio, M Incerti, D Pala, S Russo, P Chiodelli, M Rusnati, A M Cantoni, R Di Lecce, E Barocelli, S Bertoni, P Ravassard, F Manenti, L Piemonti, F Ferlenghi, A Lodola, M Tognolini
ABSTRACT
Eph/ephrin interactions and their bidirectional signaling are integral part of the complex communication system between β-cells, essential for glucose homeostasis. Indeed, Eph/ephrin system was shown to be directly involved in the glucose-stimulated insulin secretion (GSIS) process occurring in the pancreatic islets. Here we tested the Eph antagonist UniPR500 as GSIS enhancer. UniPR500 was validated as EphA5-ephrin-A5 inhibitor in vitro and its efficacy as GSIS enhancer was assessed on EndoC-βH1 cells. The selectivity of UniPR500 was evaluated by testing this compound on a panel of well-known molecular targets responsible for the regulation of glucose homeostasis. Plasmatic levels of UniPR500 were measured by HPLC/MS approach after oral administration. Finally, UniPR500 was tested as hypoglycemic agent in healthy mice, in a non-genetic mouse model of insulin resistance (IR) and in a non-genetic mouse model of type 1 diabetes (T1D). The compound is an orally bioavailable and selective Eph antagonist, able to increase GSIS from EndoC-βH1 cells. When tested in vivo UniPR500 showed to improve glucose tolerance in healthy and IR mice. As expected by a GSIS enhancer acting on healthy β-cells, UniPR500 was ineffective when tested on a non-genetic mouse model of type 1 diabetes, where pancreatic function was severely compromised. In conclusion our findings suggest that Eph targeting is a new and valuable pharmacological strategy in the search of new hypoglycemic agents.