MilliporeSigma
  • A blockade of PI3Kγ signaling effectively mitigates angiotensin II-induced renal injury and fibrosis in a mouse model.
A blockade of PI3Kγ signaling effectively mitigates angiotensin II-induced renal injury and fibrosis in a mouse model.
Scientific reports (2018-07-22)
Xinyi Yu, Yunfeng Xia, Liyi Zeng, Xi Zhang, Liqun Chen, Shujuan Yan, Ruyi Zhang, Chen Zhao, Zongyue Zeng, Yi Shu, Shifeng Huang, Jiayan Lei, Chengfu Yuan, Linghuan Zhang, Yixiao Feng, Wei Liu, Bo Huang, Bo Zhang, Wenping Luo, Xi Wang, Hongmei Zhang, Rex C Haydon, Hue H Luu, Tong-Chuan He, Hua Gan
ABSTRACT
Chronic kidney disease (CKD) poses a formidable challenge for public healthcare worldwide as vast majority of patients with CKD are also at risk of accelerated cardiovascular disease and death. Renal fibrosis is the common manifestation of CKD that usually leads to end-stage renal disease although the molecular events leading to chronic renal fibrosis and eventually chronic renal failure remain to be fully understood. Nonetheless, emerging evidence suggests that an aberrant activation of PI3Kγ signaling may play an important role in regulating profibrotic phenotypes. Here, we investigate whether a blockade of PI3Kγ signaling exerts any beneficial effect on alleviating kidney injury and renal fibrosis. Using a mouse model of angiotensin II (Ang II)-induced renal damage, we demonstrate that PI3Kγ inhibitor AS605240 effectively mitigates Ang II-induced increases in serum creatinine and blood urea nitrogen, renal interstitial collagen deposition, the accumulation of ECM proteins and the expression of α-Sma and fibrosis-related genes in vivo. Mechanistically, we reveal that AS605240 effectively inhibits Ang II-induced cell proliferation and phosphorylation of Akt in fibroblast cells. Furthermore, we demonstrate that Ang II-upregulated expression of IL-6, Tnf-α, IL-1β and Tgf-β1 is significantly attenuated in the mice treated with AS605240. Taken together, our results demonstrate that PI3Kγ may function as a critical mediator of Ang II-induced renal injury and fibrosis. It is thus conceivable that targeted inhibition of PI3Kγ signaling may constitute a novel therapeutic approach to the clinical management of renal fibrosis, renal hypertension and/or CKD.