  • Search Results
  • Bi-layered silane-TiO2/collagen coating to control biodegradation and biointegration of Mg alloys.
Bi-layered silane-TiO2/collagen coating to control biodegradation and biointegration of Mg alloys.
Materials science & engineering. C, Materials for biological applications (2018-11-15)
Laura C Córdoba, Christophe Hélary, Fátima Montemor, Thibaud Coradin
ABSTRACT
Magnesium alloys have shown high potential as biodegradable implants for bone repair applications. However, their fast degradation in physiological media demands tuning their corrosion rate to accompany the natural tissue healing processes. Here, a new bi-layered silane-TiO2/collagen coating efficient in stabilizing and biofunctionalizing the surface of AZ31 and ZE41 Mg alloys is presented. Corrosion tests performed in cell culture medium over 7 weeks showed that the bi-layered coating promotes the formation of a stable layer of Mg(OH)2/MgCO3/CaCO3 that provides effective protection to the alloys at advanced immersion stages. The intrinsic reactivity of each alloy plus formation of transitory calcium phosphate phases, resulted in distinct corrosion behavior in the short term. Cell experiments showed that the bi-layered coating improved osteoblasts and fibroblasts proliferation compared to bare and silane-TiO2-coated alloys. Different responses in terms of cell adhesion could be related to the intrinsic corrosion rate of each alloy and some toxicity from the alloying elements. The results evidenced the complex interplay between alloy nature, coating-alloy combination and cell type. The silane-TiO2/collagen coating showed to be a promising strategy to improve cell response and viability and to control degradation rate of Mg alloys in the long term.
Sigma-Aldrich
(3-Glycidyloxypropyl)trimethoxysilane, ≥98%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.