  • Search Results
  • Acquired HER2 mutations in ER+ metastatic breast cancer confer resistance to estrogen receptor-directed therapies.
Acquired HER2 mutations in ER+ metastatic breast cancer confer resistance to estrogen receptor-directed therapies.
Nature genetics (2018-12-12)
Utthara Nayar, Ofir Cohen, Christian Kapstad, Michael S Cuoco, Adrienne G Waks, Seth A Wander, Corrie Painter, Samuel Freeman, Nicole S Persky, Lori Marini, Karla Helvie, Nelly Oliver, Orit Rozenblatt-Rosen, Cynthia X Ma, Aviv Regev, Eric P Winer, Nancy U Lin, Nikhil Wagle
ABSTRACT
Seventy percent of breast cancers express the estrogen receptor (ER), and agents that target the ER are the mainstay of treatment. However, virtually all people with ER+ breast cancer develop resistance to ER-directed agents in the metastatic setting. Beyond mutations in the ER itself, which occur in 25-30% of people treated with aromatase inhibitors1-4, knowledge about clinical resistance mechanisms remains incomplete. We identified activating HER2 mutations in metastatic biopsies from eight patients with ER+ metastatic breast cancer who had developed resistance to aromatase inhibitors, tamoxifen or fulvestrant. Examination of treatment-naive primary tumors in five patients showed no evidence of pre-existing mutations in four of five patients, suggesting that these mutations were acquired under the selective pressure of ER-directed therapy. The HER2 mutations and ER mutations were mutually exclusive, suggesting a distinct mechanism of acquired resistance to ER-directed therapies. In vitro analysis confirmed that the HER2 mutations conferred estrogen independence as well as-in contrast to ER mutations-resistance to tamoxifen, fulvestrant and the CDK4 and CDK6 inhibitor palbociclib. Resistance was overcome by combining ER-directed therapy with the irreversible HER2 kinase inhibitor neratinib.
Sigma-Aldrich
Anti-phospho-erbB-2/HER-2 (Tyr1248) Antibody, Upstate®, from rabbit
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg