  • Search Results
  • A novel dimeric flavonol glycoside from Cynanchum acutum subsp. sibiricum.
A novel dimeric flavonol glycoside from Cynanchum acutum subsp. sibiricum.
Natural product research (2018-06-13)
Si-Wen Yuan, Wei Dai, Xin-Hui Pan, Yan Lu, Dao-Feng Chen, Qi Wang
ABSTRACT
A novel dimeric flavonol glycoside, Cynanflavoside A (1), together with six analogues, kaempferol-3-O-α-L-rhamnopyranosyl-(1→2)-β-D-glucopyranoside (2), quercetin-3-O-α-L-rhamnopyranosyl-(1→2)-β-D-glucopyranoside (3), kaempferol-3-O-α-L-rhamnopyranosyl-(1→2)-β-D-xylopyranoside (4), quercetin-3-O-α-L-rhamnopyranosyl-(1→2)-β-D-xylopyranoside (5), kaempferol-3-O-β-D-glucopyranosyl-7-O-α-L-rhamnopyranoside (6), and quercetin-3-O-galactoside (7) were isolated from the n-butyl alcohol extract of Cynanchum acutum subsp. sibiricum. Their structures were determined spectroscopically and compared with previously reported spectral data. All compounds were evaluated for their anti-complementary activity in vitro, and only compound 5 exhibited anti-complement effects with CH50 value of 0.33 mM.