MilliporeSigma
Locomotion-dependent remapping of distributed cortical networks.
Nature neuroscience (2019-03-13)
Kelly B Clancy, Ivana Orsolic, Thomas D Mrsic-Flogel
ABSTRACT
The interactions between neocortical areas are fluid and state-dependent, but how individual neurons couple to cortex-wide network dynamics remains poorly understood. We correlated the spiking of neurons in primary visual (V1) and retrosplenial (RSP) cortex to activity across dorsal cortex, recorded simultaneously by widefield calcium imaging. Neurons were correlated with distinct and reproducible patterns of activity across the cortical surface; while some fired predominantly with their local area, others coupled to activity in distal areas. The extent of distal coupling was predicted by how strongly neurons correlated with the local network. Changes in brain state triggered by locomotion strengthened affiliations of V1 neurons with higher visual and motor areas, while strengthening distal affiliations of RSP neurons with sensory cortices. Thus, the diverse coupling of individual neurons to cortex-wide activity patterns is restructured by running in an area-specific manner, resulting in a shift in the mode of cortical processing during locomotion.
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO