  • Search Results
  • All tubulins are not alike: heterodimer dissociation differs among different biological sources
All tubulins are not alike: heterodimer dissociation differs among different biological sources
The Journal of biological chemistry (2019)
Montecinos-Franjola F, et al.