  • Search Results
  • 3D spheroid model of mIMCD3 cells for studying ciliopathies and renal epithelial disorders
3D spheroid model of mIMCD3 cells for studying ciliopathies and renal epithelial disorders
Nature Protocols (2014)
Giles RH, et al.