  • Search Results
  • An assessment of the corrosion protection of AA2024-T3 treated with vinyltrimethoxysilane/(3-glycidyloxypropyl) trimethoxysilane.
An assessment of the corrosion protection of AA2024-T3 treated with vinyltrimethoxysilane/(3-glycidyloxypropyl) trimethoxysilane.
Corrosion Science (2015)
Dalmoro V et al.