  • Search Results
  • Organic-inorganic bonding in chitosan-silica hybrid networks: physical properties.
Organic-inorganic bonding in chitosan-silica hybrid networks: physical properties.
Journal of Polymer Science. Part B, Polymer Physics (2015)
Trujillo, et al.