  • Search Results
  • Room temperature ionic liquids for epoxy nanocomposite synthesis: Direct dispersion and cure.
Room temperature ionic liquids for epoxy nanocomposite synthesis: Direct dispersion and cure.
Composites Science and Technology (2013)
Throckmorton JA, et al.