  • Search Results
  • Effect of (3-glycidyloxypropyl) trimethoxysilane (GOPS) on the electrical properties of PEDOT: PSS films.
Effect of (3-glycidyloxypropyl) trimethoxysilane (GOPS) on the electrical properties of PEDOT: PSS films.
Journal of Polymer Science. Part B, Polymer Physics (2017)
kansson H, et al.