  • Search Results
  • Insulin tolerance test under anaesthesia to measure tissue-specific insulin-stimulated glucose disposal
Insulin tolerance test under anaesthesia to measure tissue-specific insulin-stimulated glucose disposal
eLife (2018)
Fazakerley DJ, et al.