  • Search Results
  • A localized surface plasmon resonance (LSPR) immunosensor for CRP detection using 4-chloro-1-naphtol (4-CN) precipitation
A localized surface plasmon resonance (LSPR) immunosensor for CRP detection using 4-chloro-1-naphtol (4-CN) precipitation
International Conference on Nano-Bio Sensing, Imaging, and Spectroscopy 2017 (2017)