  • Search Results
  • Metastatic canine mammary carcinomas can be identified by a gene expression profile that partly overlaps with human breast cancer profiles
Metastatic canine mammary carcinomas can be identified by a gene expression profile that partly overlaps with human breast cancer profiles
BMC Cancer (2010)
Klopfleisch R, et al.