MilliporeSigma
  • Expression of orexin A and its receptor 1 in the choroid plexuses from buffalo brain.
Expression of orexin A and its receptor 1 in the choroid plexuses from buffalo brain.
Neuropeptides (2009-03-03)
Simona Tafuri, Luigi M Pavone, Vincenzo Mastellone, Anna Spina, Luigi Avallone, Alfredo Vittoria, Norma Staiano, Gaetano Scala
ABSTRACT
The hypothalamic peptide orexin A, deriving from the proteolytic cleavage of the precursor molecule prepro-orexin, has a wide range of physiological effects including the regulation of feeding behaviour, neuroendocrine functions, sleep-wake cycle, and energy homeostasis. Lowered excretion of orexin A into the cerebrospinal fluid (CSF) plays a pathological role in animal and human narcolepsy. Altered levels of orexin A into the CSF have been also found in numerous disorders of the central nervous system, including Parkinson's and Huntington's disease, dementia, and depressive disorders. While the localization of orexin A and its receptor 1, OX(1), has been elicited in many regions of the mammalian brain and in peripheral organs, there are no information on the expression of the neuropeptide and its receptor 1 in the choroid plexuses (CPs) producing the CSF. In this study, we investigated the expression of orexin A and OX(1) in the CPs from the brain of an adult mammalian species, Bubalis bubalis, by immunogold-labelling in scanning electron microscopy. Both orexin A and OX(1) immuno-reactivity appeared to be widely distributed on the surface of choroid epithelium. Interestingly, a marked orexin A labelling was detected in the areas surrounding the CP blood capillaries. The expression of prepro-orexin and OX(1) mRNA transcripts of 200 and 300 bp, respectively, was assessed in the CPs by reverse-transcription polymerase chain reaction, while Western blotting analysis confirmed the presence of these two proteins in the tissue. Our findings provide the first evidence for orexin A and OX(1) expression in the CPs from mammalian brain, and suggest that the levels of orexin A into the CSF are probably regulated by CP activity.